Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI D-(+)-Melibiose Monohydrate, >99.0% HPLC, 1 g

Catalog No. 502781135
Encompass_Preferred

D-(+)-Melibiose Monohydrate is a high-purity disaccharide supplied as a white solid. Designed for professional research laboratory use, this glycoscience reagent is suitable for applications such as glyco-chemical analysis and prebiotic studies. It is stable under normal conditions and should be stored in a cool, dark environment at temperatures below 15°C.

  • Purity: ≥99.0% (HPLC)
  • Molecular formula: C12H22O11·H2O
  • Molecular weight: 360.32 g/mol
  • Melting point: 185°C (dec.)
  • Specific rotation: +132° to +139° (c=4, H2O)
  • Physical state: Solid

Catalog No. 50-278-1135 Supplier Grainger Supplier No. 19KR44
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Melibiose Monohydrate is a high-purity disaccharide supplied as a white solid. Designed for professional research laboratory use, this glycoscience reagent is suitable for applications such as glyco-chemical analysis and prebiotic studies. It is stable under normal conditions and should be stored in a cool, dark environment at temperatures below 15°C.

  • Purity: ≥99.0% (HPLC)
  • Molecular formula: C12H22O11·H2O
  • Molecular weight: 360.32 g/mol
  • Melting point: 185°C (dec.)
  • Specific rotation: +132° to +139° (c=4, H2O)
  • Physical state: Solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.