Learn More
Medchemexpress LLC D-melibiose | 585-99-9 | 98.4% | 342.30 | C12H22O11 | 100 MG
SDP
Catalog No. 5000240266
Encompass_Preferred
D-Melibiose (Standard) is an analytical reference standard of the disaccharide D-melibiose (CAS 585-99-9) intended for research and analytical use. The material is a white to off-white solid with molecular formula C12H22O11 and molecular weight 342.30. Recommended storage is -20 °C, sealed, protected from light and moisture.
  • Analytical reference for method development and calibration.
  • High HPLC purity (99.86% reported on COA).
  • Assay reported as 98.42% (COA), with moisture and residue specifications.
  • Solid form for accurate weighing and handling.
  • Recommended frozen storage to maintain stability.
Catalog No. 50-002-40266 Supplier Medchemexpress LLC Supplier No. HY107824R100MG
D-Melibiose (Standard) is an analytical reference standard of the disaccharide D-melibiose (CAS 585-99-9) intended for research and analytical use. The material is a white to off-white solid with molecular formula C12H22O11 and molecular weight 342.30. Recommended storage is -20 °C, sealed, protected from light and moisture.
  • Analytical reference for method development and calibration.
  • High HPLC purity (99.86% reported on COA).
  • Assay reported as 98.42% (COA), with moisture and residue specifications.
  • Solid form for accurate weighing and handling.
  • Recommended frozen storage to maintain stability.