Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-melibiose | 585-99-9 | MFCD00168065 | >=98.0% | 342.30 | C12H22O11 | 10 MG
SDP

Catalog No. 5000240267
Encompass_Preferred

D-melibiose is an analytical reference standard intended for research and analytical applications. It is a disaccharide composed of galactose and glucose linked by an alpha (1-6) glycosidic bond, supplied as a material for calibration, method development, and quantitative assays.

  • Analytical reference for calibration and method validation.
  • Suitable for chromatographic and mass spectrometry assays.
  • Disaccharide composition: galactose and glucose with alpha (1-6) glycosidic linkage.
  • Molecular formula C12H22O11; molecular weight 342.30 g/mol.
  • CAS number 585-99-9 for unambiguous identification.

Catalog No. 50-002-40267 Supplier Medchemexpress LLC Supplier No. HY107824R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-melibiose is an analytical reference standard intended for research and analytical applications. It is a disaccharide composed of galactose and glucose linked by an alpha (1-6) glycosidic bond, supplied as a material for calibration, method development, and quantitative assays.

  • Analytical reference for calibration and method validation.
  • Suitable for chromatographic and mass spectrometry assays.
  • Disaccharide composition: galactose and glucose with alpha (1-6) glycosidic linkage.
  • Molecular formula C12H22O11; molecular weight 342.30 g/mol.
  • CAS number 585-99-9 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.