Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-melibiose | 585-99-9 | MFCD00198188 | 99.8% | 342.30 g·mol-1 | C12H22O11 | 1 ML
SDP

Catalog No. 5000244478
Encompass_Preferred

D-melibiose is a disaccharide composed of one galactose and one glucose linked by an alpha (1-6) glycosidic bond. It is provided as a high-purity analytical standard and as a 10 mM solution (1 mL) in DMSO for use in biochemical assays, metabolite profiling, and microbiological applications.

  • High purity (99.8%).
  • Molecular weight 342.30 g·mol-1.
  • Chemical formula C12H22O11.
  • Available as 10 mM solution in DMSO, 1 mL.
  • Soluble in water and DMSO.
  • Suitable as a metabolite standard, microbial carbon source, and biochemical reagent.

Catalog No. 50-002-44478 Supplier Medchemexpress LLC Supplier No. HY10782410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-melibiose is a disaccharide composed of one galactose and one glucose linked by an alpha (1-6) glycosidic bond. It is provided as a high-purity analytical standard and as a 10 mM solution (1 mL) in DMSO for use in biochemical assays, metabolite profiling, and microbiological applications.

  • High purity (99.8%).
  • Molecular weight 342.30 g·mol-1.
  • Chemical formula C12H22O11.
  • Available as 10 mM solution in DMSO, 1 mL.
  • Soluble in water and DMSO.
  • Suitable as a metabolite standard, microbial carbon source, and biochemical reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.