Learn More
Medchemexpress LLC D-Melibiose 5g | 585-99-9 | 5G
SDP
Catalog No. 5000242264
Encompass_Preferred
D-Melibiose is a disaccharide which is composed of one galactose and one glucose moiety in an alpha (1-6) glycosidic linkage
Catalog No. 50-002-42264 Supplier Medchemexpress LLC Supplier No. HY1078245G
D-Melibiose is a disaccharide which is composed of one galactose and one glucose moiety in an alpha (1-6) glycosidic linkage