Learn More
Alkali Scientific D Melibiose (Monohydrate), 1 G
SDP
Catalog No. 5000660938
Encompass_Preferred
D Melibiose (Monohydrate), 1 g, is a carbohydrate reagent commonly used in laboratory research for studying sugar metabolism and enzymatic activity. Melibiose, a disaccharide made of glucose and galactose, serves as a substrate for specific enzymes such as melibiose phosphorylase. This reagent is useful in biochemical assays to investigate enzyme kinetics, metabolism, and microbial fermentation processes. It is often used in studies focused on the fermentation of sugars by certain bacterial strains, as well as in cell culture media for studying the metabolic pathways of organisms that utilize melibiose. This reagent is an essential tool in carbohydrate-based research.
Catalog No. 50-006-60938 Supplier Alkali Scientific Supplier No. DME002CP
D Melibiose (Monohydrate), 1 g, is a carbohydrate reagent commonly used in laboratory research for studying sugar metabolism and enzymatic activity. Melibiose, a disaccharide made of glucose and galactose, serves as a substrate for specific enzymes such as melibiose phosphorylase. This reagent is useful in biochemical assays to investigate enzyme kinetics, metabolism, and microbial fermentation processes. It is often used in studies focused on the fermentation of sugars by certain bacterial strains, as well as in cell culture media for studying the metabolic pathways of organisms that utilize melibiose. This reagent is an essential tool in carbohydrate-based research.