Learn More
Research Products International Corp RPI D-Melibiose Monohydrate, 1 Gram

Supplier: Research Products International Corp M224501.0
CAS Number: 585-99-9
Molecular Formula: C12H22O11 •: H2O
Molecular Weight: 360.31
Chemical Purity: >98%
Soluble in: Water
Store at: Room Temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.