Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Methionine, S-oxide | 21056-56-4 | 98.7% | 1 ML
SDP

Catalog No. 5000340593
Encompass_Preferred

D-methionine sulfoxide is the D-isomer of Methionine sulfoxide, an oxidation product of methionine. Methionine is the limiting amino acid in milk or leguminous proteins, which is easily oxidized during storage or processing. This product, appearing as a white to off-white solid, has a molecular weight of 165.21 and a chemical formula of C5H11NO3S. It is intended for research use only.

  • Purity of 98.74%
  • Solid appearance
  • White to off-white color
  • Molecular weight of 165.21
  • Chemical formula C5H11NO3S
  • Endogenous metabolite

Catalog No. 50-003-40593 Supplier Medchemexpress LLC Supplier No. HY12977010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-methionine sulfoxide is the D-isomer of Methionine sulfoxide, an oxidation product of methionine. Methionine is the limiting amino acid in milk or leguminous proteins, which is easily oxidized during storage or processing. This product, appearing as a white to off-white solid, has a molecular weight of 165.21 and a chemical formula of C5H11NO3S. It is intended for research use only.

  • Purity of 98.74%
  • Solid appearance
  • White to off-white color
  • Molecular weight of 165.21
  • Chemical formula C5H11NO3S
  • Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.