Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Methionine, S-oxide | 21056-56-4 | 98.7% | 25 MG
SDP

Catalog No. 5000342765
Encompass_Preferred

D-Methionine sulfoxide is the D-isomer of Methionine sulfoxide, an oxidation product of methionine. Methionine is recognized as the limiting amino acid in milk or leguminous proteins, which is susceptible to oxidation during storage or processing.

  • Molecular weight of 165.21
  • Molecular formula of C₅H₁₁NO₃S
  • Appears as a white to off-white solid
  • Store at -20°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)
  • Soluble in water at 125 mg/mL
  • Soluble in PBS at 50 mg/mL
  • Initial source is an endogenous metabolite

Catalog No. 50-003-42765 Supplier Medchemexpress LLC Supplier No. 25MGHY129770
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Methionine sulfoxide is the D-isomer of Methionine sulfoxide, an oxidation product of methionine. Methionine is recognized as the limiting amino acid in milk or leguminous proteins, which is susceptible to oxidation during storage or processing.

  • Molecular weight of 165.21
  • Molecular formula of C₅H₁₁NO₃S
  • Appears as a white to off-white solid
  • Store at -20°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)
  • Soluble in water at 125 mg/mL
  • Soluble in PBS at 50 mg/mL
  • Initial source is an endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.