Learn More
Medchemexpress LLC D-methionine sulfoxide hydrochloride | 00-00-0 | MFCD18828262 | 99.8% | C5H12ClNO3S | 10MG
SDP
Catalog No. 5000261551
Encompass_Preferred
D-methionine sulfoxide hydrochloride is the D-isomer of methionine sulfoxide, an oxidation product of methionine. Supplied as a hydrochloride salt in small milligram-scale quantities, the compound is used in biochemical and analytical research, including studies of oxidative stress and methionine sulfoxide reduction.
Catalog No. 50-002-61551 Supplier Medchemexpress LLC Supplier No. HY129770A10MG
D-methionine sulfoxide hydrochloride is the D-isomer of methionine sulfoxide, an oxidation product of methionine. Supplied as a hydrochloride salt in small milligram-scale quantities, the compound is used in biochemical and analytical research, including studies of oxidative stress and methionine sulfoxide reduction.