Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Methionine sulfoxide (hydrochloride) | 100 MG
SDP

Catalog No. 5000338956
Encompass_Preferred

D-Methionine sulfoxide hydrochloride is a chemical compound with the formula C5H12ClNO3S and a molecular weight of 201.67. This product is identified for use as a laboratory chemical and for the manufacture of substances, appearing as a solid. It demonstrates chemical stability under recommended storage conditions, emphasizing safety in handling and storage.

  • Chemical formula: C5H12ClNO3S
  • Molecular weight: 201.67
  • Appears as a solid
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture and light
  • Can be shipped at room temperature if transit is less than two weeks
  • Recommended for laboratory use and substance manufacturing

Catalog No. 50-003-38956 Supplier Medchemexpress LLC Supplier No. HY129770A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Methionine sulfoxide hydrochloride is a chemical compound with the formula C5H12ClNO3S and a molecular weight of 201.67. This product is identified for use as a laboratory chemical and for the manufacture of substances, appearing as a solid. It demonstrates chemical stability under recommended storage conditions, emphasizing safety in handling and storage.

  • Chemical formula: C5H12ClNO3S
  • Molecular weight: 201.67
  • Appears as a solid
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture and light
  • Can be shipped at room temperature if transit is less than two weeks
  • Recommended for laboratory use and substance manufacturing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.