Learn More
Cayman Chemical D-Mnoheptulose 10mg
SDP
Catalog No. 502967574
Encompass_Preferred
A heptose that inhibits glucokinases and hexokinases from diverse organisms through competition with D-glucose (Ki = 0.25 mM); blocks glucose oxidation and glucose-mediated insulin release from pancreatic islet cells
Catalog No. 50-296-7574 Supplier Cayman Chemical Supplier No. 1654810 MG
A heptose that inhibits glucokinases and hexokinases from diverse organisms through competition with D-glucose (Ki = 0.25 mM); blocks glucose oxidation and glucose-mediated insulin release from pancreatic islet cells