Learn More
Cayman Chemical D-Mannoheptulose, Purity: ≥98%, CAS: 3615-44-9, 50 mg
SDP
Catalog No. 502968385
Encompass_Preferred
A heptose that inhibits glucokinases and hexokinases from diverse organisms through competition with D-glucose (Ki = 0.25 mM); blocks glucose oxidation and glucose-mediated insulin release from pancreatic islet cells
Catalog No. 50-296-8385 Supplier Cayman Chemical Supplier No. 1654850 MG
A heptose that inhibits glucokinases and hexokinases from diverse organisms through competition with D-glucose (Ki = 0.25 mM); blocks glucose oxidation and glucose-mediated insulin release from pancreatic islet cells