Learn More
Cayman Chemical D-Mnose-6-PhosphatesodIum 10mg
SDP
Catalog No. 502982727
Encompass_Preferred
A phosphorylated aldohexose that can be used to produce glucose, mannitol, or N-glycans; also a ligand for both cation-dependent and -independent receptors and binds, as part of an N-glycan, to MRH domains on other proteins
Catalog No. 50-298-2727 Supplier Cayman Chemical Supplier No. 1889010 MG
A phosphorylated aldohexose that can be used to produce glucose, mannitol, or N-glycans; also a ligand for both cation-dependent and -independent receptors and binds, as part of an N-glycan, to MRH domains on other proteins