Learn More
Medchemexpress LLC D-moses (D-45) | 2084850-77-9 | 360.46 | C21H24N6 | 5 MG
SDP
Catalog No. 5000211445
Encompass_Preferred
D-Moses is the enantiomer of a PCAF bromodomain inhibitor and is supplied as an inactive control compound for use in biochemical and cell-based assays. It is provided as a solid and intended for use as a negative control in experimental workflows.
  • Serves as an inactive control for PCAF bromodomain assays.
  • Suitable for biochemical and cell-based assay validation.
  • Supplied as a solid with recommended cold storage for stability.
  • Technical documentation includes molecular formula and molecular weight.
  • Pack sizes are appropriate for screening and validation experiments.
Catalog No. 50-002-11445 Supplier Medchemexpress LLC Supplier No. HY101125B5MG
D-Moses is the enantiomer of a PCAF bromodomain inhibitor and is supplied as an inactive control compound for use in biochemical and cell-based assays. It is provided as a solid and intended for use as a negative control in experimental workflows.
  • Serves as an inactive control for PCAF bromodomain assays.
  • Suitable for biochemical and cell-based assay validation.
  • Supplied as a solid with recommended cold storage for stability.
  • Technical documentation includes molecular formula and molecular weight.
  • Pack sizes are appropriate for screening and validation experiments.