Learn More
Medchemexpress LLC D-Moses (D-45) | 2084850-77-9 | >98.0% | 360.46 g/mol | C21H24N6 | 10 MG
SDP
Catalog No. 5000213520
Encompass_Preferred
D-Moses (D-45) is the D-enantiomer of L-Moses and is provided as an inactive control compound for studies of the p300/CBP-associated factor (PCAF) bromodomain. It is intended for research use and is supplied as a powder with reported high purity for biochemical and cellular assays (CAS 2084850-77-9).
  • Inactive enantiomer control for bromodomain assays.
  • Suitable for biochemical and cell-based experiments.
  • Reported purity greater than 98%.
  • Available in small, precise milligram sizes for screening.
  • Powder form with limited aqueous solubility; soluble in common organic solvents.
Catalog No. 50-002-13520 Supplier Medchemexpress LLC Supplier No. HY101125B10MG
D-Moses (D-45) is the D-enantiomer of L-Moses and is provided as an inactive control compound for studies of the p300/CBP-associated factor (PCAF) bromodomain. It is intended for research use and is supplied as a powder with reported high purity for biochemical and cellular assays (CAS 2084850-77-9).
  • Inactive enantiomer control for bromodomain assays.
  • Suitable for biochemical and cell-based experiments.
  • Reported purity greater than 98%.
  • Available in small, precise milligram sizes for screening.
  • Powder form with limited aqueous solubility; soluble in common organic solvents.