Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-myo-Inositol-1,2,4,5-tetrakis(phosphate) sodium salt, Thermo Scientific Chemicals

Catalog No. AAJ66138EXZ
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66138EXZ 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66138EXZ Supplier Thermo Scientific Chemicals Supplier No. J66138EXZ
Add to Cart

 

A useful IP3R activator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD12912299
Synonym 1,2,4,5-IP4 sodium salt; Ins(1,2,4,5)-P4 sodium salt

Specifications

Color White
Quantity 0.1 mg
Solubility Information Soluble in water
Formula Weight 588
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-1,2,4,5-tetrakis(phosphate) sodium salt
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.