Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN19468890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19468890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19468890 Supplier MP Biomedicals, Inc Supplier No. 0219468890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Color White
Density 6.2g/mL
Assay Percent Range 99.26%
Quantity 500 g
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material D-Myo-inositol, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.