Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-myo-Inositol, MP Biomedicals™

Catalog No. MP210205291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210205291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210205291 Supplier MP Biomedicals, Inc Supplier No. 0210205291
Add to Cart
Add to Cart

Plays an important role as a structural element for secondary messengers in eukaryotic cells. D-myo-Inositol, MP Biomedicals is a growth factor for animals and microorganisms. It serves as an important component of phosphatidylinositol (PI) and the phosph

Myo-Inositol is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells.

  • It is a component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers
  • Inositol or cyclohexane-1,2,3,4,5,6-hexol is a chemical compound, a sixfold alcohol (polyol) of cyclohexane
  • It exists in nine possible stereoisomers, of which the most prominent form, widely occurring in nature, is cis-1,2,3,5-trans-4,6-cyclohexanehexol, or myo-inositol (former name meso-inositol)
  • Inositol is a carbohydrate, though not a classical sugar. It has a taste which has been assayed at half the sweetness of table sugar (sucrose)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 220°C to 230°C
pH Neutral to Litmus (10% aq. soln.)
Quantity 1 kg
Solubility Information Readily soluble in water (50mg/mL); slightly soluble in alcohol; insoluble in ether and other common organic solvents.
Formula Weight 180.2
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.