Learn More
Enzo Life Sciences D-myo-Inositol 1,4,5-trisphosphate . hexapotassium salt (5mg). CAS: 103476-24-0
SDP
Catalog No. 502008669
Encompass_Preferred
Product of phosphatidylinositol phospholipid hydrolysis. Causes release of calcium from intracellular stores. Alternative name: Ins(1,4,5)P3 . 6K, IP3. Purity: ≥98% (TLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8669 Supplier Enzo Life Sciences Supplier No. ALX307007M005
Product of phosphatidylinositol phospholipid hydrolysis. Causes release of calcium from intracellular stores. Alternative name: Ins(1,4,5)P3 . 6K, IP3. Purity: ≥98% (TLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C.