Learn More
Medchemexpress LLC D-N-Acetylgalactosamine-13C | 478518-53-5 | 99.95% | 222.20 | 1 MG
SDP
Catalog No. 5000403617
Encompass_Preferred
D-N-Acetylgalactosamine-13C is the 13C labeled D-N-Acetylgalactosamine. D-N-Acetylgalactosamine is an endogenous metabolite. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope
Catalog No. 50-004-03617 Supplier Medchemexpress LLC Supplier No. HY33212S1MG
D-N-Acetylgalactosamine-13C is the 13C labeled D-N-Acetylgalactosamine. D-N-Acetylgalactosamine is an endogenous metabolite. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope