Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ D-Net with Expandable Pole
SDPSCI_ED

Catalog No. p-2836215 Shop All LaMotte Company Products
Change view
Click to view available options
Product Type:
D-net with Expandable Pole
Replacement Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S85027 D-net with Expandable Pole
S85027B Replacement Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S85027 Supplier LaMotte™ Supplier No. 0168
Add to Cart
Edge
Add to Cart

Bag attached to metal frame with clamp rings.

  • 12 in. (30.5cm) rim dip net
  • Rim fitted with heavy canvas bag, approximately 6.5 in. deep (16.5cm)
  • Bag bottom made of approximately 500μm nylon mesh
  • Canvas skirt extends 3 in. (7.6cm) below bag bottom to protect mesh
  • Aluminum pole extends from 52 to 84 in. (137.2 to 213.3cm)

Specifications

Product Type D-net with Expandable Pole
For Use With (Application) Environmental education
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.