Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Nipecotic acid, 96+%

Catalog No. AAH5743403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5743403 1 g
AAH5743406 5 g
AAH5743414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5743403 Supplier Thermo Scientific Chemicals Supplier No. H5743403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 25137-00-2 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25137-00-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD01630787
InChI Key XJLSEXAGTJCILF-RXMQYKEDSA-N
Synonym r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
PubChem CID 1796510
ChEBI CHEBI:221278
IUPAC Name (3R)-piperidine-3-carboxylic acid
SMILES C1CC(CNC1)C(=O)O

Specifications

Melting Point 250°C to 255°C
Quantity 1 g
Optical Rotation −5° (c=1 in Water)
Formula Weight 129.16
Percent Purity ≥96%
Chemical Name or Material D-Nipecotic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber TM6125390
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.