Learn More
Medchemexpress LLC D-nmappd ((1R,2R)-B13) | 35922-06-6 | MFCD08062172 | 99.7% | 422.56 g/mol | C23H38N2O5 | 5 MG
SDP
Catalog No. 5000264841
Encompass_Preferred
D-NMAPPD is an acid ceramidase inhibitor that regulates NMDA receptor properties by enhancing endogenous ceramide production. It has been reported to induce apoptosis and exhibit anticancer effects, and is supplied as a high-purity small-molecule reagent for research use.
  • Acid ceramidase inhibitor used in biochemical and cellular studies.
  • Regulates NMDA receptor properties via enhanced ceramide production.
  • Reported to induce apoptosis and show anticancer activity in vitro.
  • Chemical formula C23H38N2O5; molecular weight 422.56 g/mol.
  • High purity: 99.7%.
  • Available in milligram quantities and DMSO solutions for research use.
Catalog No. 50-002-64841 Supplier Medchemexpress LLC Supplier No. HY1249625MG
D-NMAPPD is an acid ceramidase inhibitor that regulates NMDA receptor properties by enhancing endogenous ceramide production. It has been reported to induce apoptosis and exhibit anticancer effects, and is supplied as a high-purity small-molecule reagent for research use.
  • Acid ceramidase inhibitor used in biochemical and cellular studies.
  • Regulates NMDA receptor properties via enhanced ceramide production.
  • Reported to induce apoptosis and show anticancer activity in vitro.
  • Chemical formula C23H38N2O5; molecular weight 422.56 g/mol.
  • High purity: 99.7%.
  • Available in milligram quantities and DMSO solutions for research use.