Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-NMAPPD | 35922-06-6 | MFCD08062172 | 99.7% | 422.56 g/mol | C23H38N2O5 | 10MM 1ML
SDP

Catalog No. 5000235264
Encompass_Preferred

D-NMAPPD ((1R,2R)-B13) is a research-grade small molecule that inhibits acid ceramidase and modulates NMDA receptor function by increasing endogenous ceramide levels. It is supplied for in vitro pharmacological and cell biology studies, including apoptosis and anticancer research. Available as solution and solid formats for assay development.

  • Acts as an acid ceramidase inhibitor.
  • Modulates NMDA receptor properties by enhancing ceramide production.
  • Available as 10 mM solution in DMSO and multiple solid sizes for flexible dosing.
  • High purity (99.74% by HPLC).
  • Molecular weight 422.56 g/mol; formula C23H38N2O5.
  • Intended for in vitro research use only.

Catalog No. 50-002-35264 Supplier Medchemexpress LLC Supplier No. HY12496210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-NMAPPD ((1R,2R)-B13) is a research-grade small molecule that inhibits acid ceramidase and modulates NMDA receptor function by increasing endogenous ceramide levels. It is supplied for in vitro pharmacological and cell biology studies, including apoptosis and anticancer research. Available as solution and solid formats for assay development.

  • Acts as an acid ceramidase inhibitor.
  • Modulates NMDA receptor properties by enhancing ceramide production.
  • Available as 10 mM solution in DMSO and multiple solid sizes for flexible dosing.
  • High purity (99.74% by HPLC).
  • Molecular weight 422.56 g/mol; formula C23H38N2O5.
  • Intended for in vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.