Learn More
Medchemexpress LLC D-Nmappd | 35922-06-6 | MFCD08062172 | 99.7% | 422.6 g/mol | C23H38N2O5 | 25MG
SDP
Catalog No. 5000238195
Encompass_Preferred
D-NMAPPD ((1R,2R)-B13) is a research-grade small-molecule acid ceramidase inhibitor that modulates NMDA receptor properties by enhancing endogenous ceramide production. It is used in biochemical and neurological research and has been reported to show anticancer activity in experimental studies. The compound is supplied in milligram-scale quantities with high reported purity suitable for laboratory applications.
  • Inhibits acid ceramidase activity.
  • Modulates NMDA receptor properties by enhancing endogenous ceramide production.
  • Reported anticancer activity in experimental studies.
  • High purity (~99.7%) suitable for research use.
  • Molecular formula C23H38N2O5; molecular weight 422.6 g/mol.
  • Available in milligram-scale package sizes for laboratory experiments.
Catalog No. 50-002-38195 Supplier Medchemexpress LLC Supplier No. HY12496225MG
D-NMAPPD ((1R,2R)-B13) is a research-grade small-molecule acid ceramidase inhibitor that modulates NMDA receptor properties by enhancing endogenous ceramide production. It is used in biochemical and neurological research and has been reported to show anticancer activity in experimental studies. The compound is supplied in milligram-scale quantities with high reported purity suitable for laboratory applications.
  • Inhibits acid ceramidase activity.
  • Modulates NMDA receptor properties by enhancing endogenous ceramide production.
  • Reported anticancer activity in experimental studies.
  • High purity (~99.7%) suitable for research use.
  • Molecular formula C23H38N2O5; molecular weight 422.6 g/mol.
  • Available in milligram-scale package sizes for laboratory experiments.