Learn More
Enzo Life Sciences D-NMMA . monoacetate (25mg). CAS: 137694-75-8
SDP
Catalog No. 502007732
Encompass_Preferred
D-NMMA is inactive as an inhibitor of NOS and may be used to explore non-specific effects of L-NMMA. Alternative name: NG-Monomethyl-D-arginine . monoacetate. Formula: C7H16N4O2 . CH3COOH. MW: 188.2 . 60.1. Purity: ≥99%. Appearance: White solid. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-7732 Supplier Enzo Life Sciences Supplier No. ALX106002M025
D-NMMA is inactive as an inhibitor of NOS and may be used to explore non-specific effects of L-NMMA. Alternative name: NG-Monomethyl-D-arginine . monoacetate. Formula: C7H16N4O2 . CH3COOH. MW: 188.2 . 60.1. Purity: ≥99%. Appearance: White solid. Solubility: Soluble in water. Long Term Storage: +4°C.