Learn More
AdipoGen D-NMMA . monoacetate (5 mg)
SDP
Supplier:  AdipoGen AGCR13580M005
Encompass_Preferred
Chemical. CAS: 137694-75-8. Formula: C7H16N4O2 . CH3COOH. MW: 188.2 . 60.1. Cell permeable control compound for L-NMMA (Prod. No. AG-CR1-3578). Inactive, shows no NOS inhibitor activity. Useful tool to investigate non-specific L-NMMA activity.
Catalog No. 50-205-3946