Learn More
Abcam D-NMMA monoacetate, L-NMMA control compound, 5MG
Catalog No. 502705889
Encompass_Preferred
MW 248.28 Da, Purity >98%. Cell-permeable control compound for L-NMMA (ab120137). Inactive, shows no NOS inhibitor activity. Useful tool to investigate non-specific L-NMMA activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5889 Supplier Abcam Supplier No. AB1443155MG
MW 248.28 Da, Purity >98%. Cell-permeable control compound for L-NMMA (ab120137). Inactive, shows no NOS inhibitor activity. Useful tool to investigate non-specific L-NMMA activity.
The product is subject to the following: Abcam Restricted Use Statement