Learn More
Cayman Chemical D-Neopterin, Purity: ≥98%, CAS: 2009-64-5, 100 mg
SDP
Catalog No. 502953004
Encompass_Preferred
An intermediate product in the synthetic pathway of biopterin that is released from macrophages upon immune stimulation with interferon-γ from activated T cells; routinely used as a biochemical tool to screen for and monitor the progression of certain diseases accompanied by hyperimmune stimulation
Catalog No. 50-295-3004 Supplier Cayman Chemical Supplier No. 12057100 MG
An intermediate product in the synthetic pathway of biopterin that is released from macrophages upon immune stimulation with interferon-γ from activated T cells; routinely used as a biochemical tool to screen for and monitor the progression of certain diseases accompanied by hyperimmune stimulation