Learn More
Cayman Chemical D-noptern 10mg
SDP
Catalog No. 502954344
Encompass_Preferred
An intermediate product in the synthetic pathway of biopterin that is released from macrophages upon immune stimulation with interferon-γ from activated T cells; routinely used as a biochemical tool to screen for and monitor the progression of certain diseases accompanied by hyperimmune stimulation
Catalog No. 50-295-4344 Supplier Cayman Chemical Supplier No. 1205710 MG
An intermediate product in the synthetic pathway of biopterin that is released from macrophages upon immune stimulation with interferon-γ from activated T cells; routinely used as a biochemical tool to screen for and monitor the progression of certain diseases accompanied by hyperimmune stimulation