Learn More
Cayman Chemical D-Neopterin, Purity: ≥98%, CAS: 2009-64-5, 50 mg
SDP
Catalog No. 502954122
Encompass_Preferred
An intermediate product in the synthetic pathway of biopterin that is released from macrophages upon immune stimulation with interferon-γ from activated T cells; routinely used as a biochemical tool to screen for and monitor the progression of certain diseases accompanied by hyperimmune stimulation
Catalog No. 50-295-4122 Supplier Cayman Chemical Supplier No. 1205750 MG
An intermediate product in the synthetic pathway of biopterin that is released from macrophages upon immune stimulation with interferon-γ from activated T cells; routinely used as a biochemical tool to screen for and monitor the progression of certain diseases accompanied by hyperimmune stimulation