Learn More
Broadley James Corporation Cartridge Kit for 19 and 25 mm DO Sensors
Catalog No. NC9762254
Encompass
Four Cartridge Kit, 25mm, Includes: One 25mL Bottle of Electrolyte, Four Membrane Cartridges, Four Sets of Internal O-Rings
Catalog No. NC9762254 Supplier Broadley James Corporation Supplier No. KA2504
Four Cartridge Kit, 25mm, Includes: One 25mL Bottle of Electrolyte, Four Membrane Cartridges, Four Sets of Internal O-Rings