Learn More
Cayman Chemical D-Ornithine lactam, Purity: ≥95%, CAS: 88763-76-2, 50 mg
SDP
Catalog No. 502992053
Encompass_Preferred
A building block; has been used in the synthesis of HCV NS5B inhibitors, as well as αVβ3 integrin receptor antagonists
Catalog No. 50-299-2053 Supplier Cayman Chemical Supplier No. 3168050 MG
A building block; has been used in the synthesis of HCV NS5B inhibitors, as well as αVβ3 integrin receptor antagonists