Learn More
Medchemexpress LLC D-Ornithine hydrochloride | 16682-12-5 | 98.0% | 168.62 | 1 ML
SDP
Catalog No. 5000403726
Encompass_Preferred
D-Ornithine hydrochloride is a key constituent of bacterial cell walls. This compound plays a role in promoting the production of L-arginine and enhancing the expression of pyrrolysine-containing proteins.
  • Constituent of bacterial cell wall.
  • Promotes production of L-arginine.
  • Enhances expression of pyrrolysine-containing proteins.
  • Selectively up-regulates L-arginine in tobacco cells.
  • Promotes insertion of pyrroline-carboxy-lysine into proteins in E. coli and HEK293 cells.
  • Stimulates production of spermidine and spermine in tobacco cells.
Catalog No. 50-004-03726 Supplier Medchemexpress LLC Supplier No. HY3451610MM/1ML
D-Ornithine hydrochloride is a key constituent of bacterial cell walls. This compound plays a role in promoting the production of L-arginine and enhancing the expression of pyrrolysine-containing proteins.
  • Constituent of bacterial cell wall.
  • Promotes production of L-arginine.
  • Enhances expression of pyrrolysine-containing proteins.
  • Selectively up-regulates L-arginine in tobacco cells.
  • Promotes insertion of pyrroline-carboxy-lysine into proteins in E. coli and HEK293 cells.
  • Stimulates production of spermidine and spermine in tobacco cells.