Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Ornithine hydrochloride | 16682-12-5 | 98.0% | 168.62 | 25 G
SDP

Catalog No. 5000402388
Encompass_Preferred

D-Ornithine hydrochloride is a constituent of bacterial cell wall. It promotes the production of L-arginine and enhances the expression of pyrrolysine-containing proteins. It is intended for research use only.

  • Constituent of bacterial cell wall
  • Promotes the production of L-arginine
  • Enhances the expression of pyrrolysine-containing proteins
  • Appearance: solid
  • Color: white to off-white
  • Initial source: endogenous metabolite
  • Results in a selective up-regulation of L-arginine in tobacco cells (1-10 mM, 24 h)
  • Promotes the insertion of pyrroline-carboxy-lysine (Pcl) into proteins in E. coli and HEK293 cells containing the pyl genes (5 mM)
  • Can stimulate production of spermidine and spermine in tobacco cells (1-10 mM)

Catalog No. 50-004-02388 Supplier Medchemexpress LLC Supplier No. 25GHY34516
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Ornithine hydrochloride is a constituent of bacterial cell wall. It promotes the production of L-arginine and enhances the expression of pyrrolysine-containing proteins. It is intended for research use only.

  • Constituent of bacterial cell wall
  • Promotes the production of L-arginine
  • Enhances the expression of pyrrolysine-containing proteins
  • Appearance: solid
  • Color: white to off-white
  • Initial source: endogenous metabolite
  • Results in a selective up-regulation of L-arginine in tobacco cells (1-10 mM, 24 h)
  • Promotes the insertion of pyrroline-carboxy-lysine (Pcl) into proteins in E. coli and HEK293 cells containing the pyl genes (5 mM)
  • Can stimulate production of spermidine and spermine in tobacco cells (1-10 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.