Learn More
Medchemexpress LLC D-ornithine, N5-(aminocarbonyl)- | 13594-51-9 | MFCD00216469 | 98.0% | 175.19 | C6H13N3O3 | 10 G
SDP
Catalog No. 5000303409
Encompass_Preferred
D-Citrulline is the D-stereoisomer of the amino acid citrulline supplied as a research-grade powder for biochemical and metabolic studies. It is used as an endogenous metabolite probe in investigations of arginine-citrulline metabolism and nitric oxide pathways, and is provided with documented purity and storage recommendations.
Catalog No. 50-003-03409 Supplier Medchemexpress LLC Supplier No. HYW01673310G
D-Citrulline is the D-stereoisomer of the amino acid citrulline supplied as a research-grade powder for biochemical and metabolic studies. It is used as an endogenous metabolite probe in investigations of arginine-citrulline metabolism and nitric oxide pathways, and is provided with documented purity and storage recommendations.