Learn More
Medchemexpress LLC HY-P1159A 5mg Medchemexpress, [D-p-Cl-Phe6,Leu17]-VIP (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1159A5MG
Encompass_Preferred
Medchemexpress, HY-P1159A 5mg [D-p-Cl-Phe6,Leu17]-VIP (TFA) CAS: [D-p-Cl-Phe6,Leu17]-VIP TFA is a competitive and selective antagonist of vasoactive intestinal peptide (VIP) receptor, with the IC50 of 125.8 nM. [D-p-Cl-Phe6,Leu17]-VIP TFA has no activity on glucagon, secretin or GRF receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3041