Learn More
Apexbio Technology LLC D-Pantethine 16816-67-4 25mg
SDP
Catalog No. 5000088386
Encompass_Preferred
D-Pantethine (16816-67-4) is a biologically-derived compound that functions as a precursor to coenzyme A via disulfide bond formation between two pantetheine molecules It is designed to participate directly in intermediary metabolic pathways including lipid and carbohydrate metabolism fatty acid beta-oxidation pyruvate processing and amino acid catabolism through its coenzyme A-related mechanisms D-Pantethine exerts its biological activity primarily through modulation of metabolic pathways Experimental studies typically employ cell-based assays or animal models examining enzyme activity and receptor modulation with reported IC50 values in the micromolar to submillimolar range depending on specific assay conditions and biological systems Based on these pharmacological properties D-Pantethine holds research potential in lipid-lowering effects anti-inflammatory processes neuroprotection and antiviral activity assessment
Catalog No. 50-000-88386 Supplier Apexbio Technology LLC Supplier No. C651125
D-Pantethine (16816-67-4) is a biologically-derived compound that functions as a precursor to coenzyme A via disulfide bond formation between two pantetheine molecules It is designed to participate directly in intermediary metabolic pathways including lipid and carbohydrate metabolism fatty acid beta-oxidation pyruvate processing and amino acid catabolism through its coenzyme A-related mechanisms D-Pantethine exerts its biological activity primarily through modulation of metabolic pathways Experimental studies typically employ cell-based assays or animal models examining enzyme activity and receptor modulation with reported IC50 values in the micromolar to submillimolar range depending on specific assay conditions and biological systems Based on these pharmacological properties D-Pantethine holds research potential in lipid-lowering effects anti-inflammatory processes neuroprotection and antiviral activity assessment