Learn More
Medchemexpress LLC D-Panthenol (standard) | 81-13-0 | 95.6% | 250 MG
SDP
Catalog No. 5000414227
Encompass_Preferred
D-Panthenol (Standard) is the analytical standard of D-Panthenol. This product is intended for research and analytical applications. D-Panthenol is the biologically-active alcohol of pantothenic acid, which leads to an elevation in the amount of coenzyme A in the cell. D-panthenol exhibits nephroprotective effect in AKI, promotes tissue repair and regeneration.
  • Intended for research and analytical applications.
  • It is the biologically-active alcohol of pantothenic acid.
  • Leads to an elevation in the amount of coenzyme A in the cell.
  • Exhibits nephroprotective effect in AKI.
  • Promotes tissue repair and regeneration.
Catalog No. 50-004-14227 Supplier Medchemexpress LLC Supplier No. 250MGHYB1391R
D-Panthenol (Standard) is the analytical standard of D-Panthenol. This product is intended for research and analytical applications. D-Panthenol is the biologically-active alcohol of pantothenic acid, which leads to an elevation in the amount of coenzyme A in the cell. D-panthenol exhibits nephroprotective effect in AKI, promotes tissue repair and regeneration.
  • Intended for research and analytical applications.
  • It is the biologically-active alcohol of pantothenic acid.
  • Leads to an elevation in the amount of coenzyme A in the cell.
  • Exhibits nephroprotective effect in AKI.
  • Promotes tissue repair and regeneration.