Learn More
Medchemexpress LLC D-(-)-Pantolactone | 599-04-2 | 100.0% | 130.14 | 10 G
SDP
Catalog No. 5000453788
Encompass_Preferred
D-(-)-Pantolactone is a significant intermediate used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It also serves as a calibrant for circular dichroism (CD).
  • Used as an intermediate in the synthesis of active compounds like the molluscicide Cyanolide A.
  • Functions as a calibrant for circular dichroism (CD).
  • Derivatives have shown good inhibitory activities against FAS and lipid accumulation, suggesting potential use in anti-obesity drug research.
Catalog No. 50-004-53788 Supplier Medchemexpress LLC Supplier No. HYW01039610G
D-(-)-Pantolactone is a significant intermediate used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It also serves as a calibrant for circular dichroism (CD).
  • Used as an intermediate in the synthesis of active compounds like the molluscicide Cyanolide A.
  • Functions as a calibrant for circular dichroism (CD).
  • Derivatives have shown good inhibitory activities against FAS and lipid accumulation, suggesting potential use in anti-obesity drug research.