Learn More
Medchemexpress LLC D-(-)-Pantolactone | 599-04-2 | 100.0% | 130.14 | 10 MM * 1 ML
SDP
Catalog No. 5000367051
Encompass_Preferred
D-(-)-Pantolactone is an important intermediate that can be used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It is also a calibrant for circular dichroism (CD). Its derivatives show good inhibitory activities against FAS and lipid accumulation, making them potentially useful in the research of anti-obesity drugs.
  • Derivatives show good inhibitory activities against FAS and lipid accumulation
  • Potentially useful in the research of anti-obesity drugs
Catalog No. 50-003-67051 Supplier Medchemexpress LLC Supplier No. HYW01039610MM/1ML
D-(-)-Pantolactone is an important intermediate that can be used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It is also a calibrant for circular dichroism (CD). Its derivatives show good inhibitory activities against FAS and lipid accumulation, making them potentially useful in the research of anti-obesity drugs.
  • Derivatives show good inhibitory activities against FAS and lipid accumulation
  • Potentially useful in the research of anti-obesity drugs