Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(-)-Pantolactone | 599-04-2 | 99.96% | 130.14 | 100 G
SDP

Catalog No. 5000453760
Encompass_Preferred

D-(-)-Pantolactone is an important intermediate used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It also functions as a calibrant for circular dichroism (CD). Derivatives of D-(-)-Pantolactone have shown good inhibitory activities against FAS and lipid accumulation, making them useful in the research of anti-obesity drugs.

  • High purity: 99.96%
  • White to off-white solid appearance
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-53760 Supplier Medchemexpress LLC Supplier No. HYW010396100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(-)-Pantolactone is an important intermediate used in the synthesis of other active compounds, such as the molluscicide Cyanolide A. It also functions as a calibrant for circular dichroism (CD). Derivatives of D-(-)-Pantolactone have shown good inhibitory activities against FAS and lipid accumulation, making them useful in the research of anti-obesity drugs.

  • High purity: 99.96%
  • White to off-white solid appearance
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.