Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Pantolactone 98.0+%, TCI America™
SDP

Catalog No. P001125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P001125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P001125G Supplier TCI America Supplier No. P001125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 599-04-2
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00005392
InChI Key SERHXTVXHNVDKA-BYPYZUCNSA-N
Synonym d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
PubChem CID 439368
ChEBI CHEBI:16719
IUPAC Name (3R)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Formula Weight 130.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Pantolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.