Learn More
Apexbio Technology LLC D-Pantothenate sodium 867-81-2 100mg
SDP
Catalog No. 5000090424
Encompass_Preferred
D-Pantothenate sodium (CAS 867-81-2) is a water-soluble small molecule that serves as a biosynthetic precursor to coenzyme A (CoA) Through its conversion to CoA D-pantothenate sodium participates in vital cellular processes involving the metabolism of carbohydrates lipids proteins and nucleic acids It modulates metabolic pathways by influencing the activity of enzymes and associated signaling networks thereby contributing to cellular metabolic regulation This compound is frequently employed in studies investigating cellular metabolism disease models related to metabolic dysfunction and the biochemical mechanisms underlying enzyme-catalyzed reactions
Catalog No. 50-000-90424 Supplier Apexbio Technology LLC Supplier No. C7117100
D-Pantothenate sodium (CAS 867-81-2) is a water-soluble small molecule that serves as a biosynthetic precursor to coenzyme A (CoA) Through its conversion to CoA D-pantothenate sodium participates in vital cellular processes involving the metabolism of carbohydrates lipids proteins and nucleic acids It modulates metabolic pathways by influencing the activity of enzymes and associated signaling networks thereby contributing to cellular metabolic regulation This compound is frequently employed in studies investigating cellular metabolism disease models related to metabolic dysfunction and the biochemical mechanisms underlying enzyme-catalyzed reactions