Learn More
Medchemexpress LLC D-Pantothenic acid-13C3,15N hemicalcium hemihydrate | 2483830-09-5 | 99.7% | C9H17NO5·1/2Ca·1/2H2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY144432S1MG
Encompass_Preferred
D-Pantothenic acid-13C3,15N hemicalcium hemihydrate is a Vitamin B5 hemicalcium hemihydrate labeled with 13C and 15N isotopes, primarily used for research. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Useful for studying the impact of isotopic labeling on pharmacokinetic and metabolic profiles
Catalog No. 50-003-54191