Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantothenic acid calcium salt, Cell culture reagent grade, MP Biomedicals™

Catalog No. ICN19472190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19472190 500 g
ICN19472180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19472190 Supplier MP Biomedicals, Inc Supplier No. 0219472190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2219-30-9
Molecular Formula C5H12ClNO2S
Molecular Weight (g/mol) 185.666
InChI Key CZDHUFYOXKHLME-DFWYDOINSA-N
Synonym d-penicillamine hydrochloride, penicillamine hydrochloride, distamine, unii-5n9jk529zj, usaf el-23, usaf a-1705, d---penicillamine hydrochloride, d-valine, 3-mercapto-, hydrochloride, valine, 3-mercapto-, hydrochloride, d
PubChem CID 92173
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid;hydrochloride
SMILES CC(C)(C(C(=O)O)N)S.Cl

Specifications

Quantity 500 g
Formula Weight 238.3
Grade Cell Culture Reagent
Density 0.5g/cm3
Chemical Name or Material D-Pantothenic acid
EINECSNumber 205-278-9
Recommended Storage Store at 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.