Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantothenic acid calcium salt, 98%

Catalog No. p-7023250
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1660922 100 g
AAA1660936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1660922 Supplier Thermo Scientific Chemicals Supplier No. A1660922
Only null left
Add to Cart

CAS: 137-08-6 | C18H34CaN2O11 | 494.551 g/mol

Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

Solubility
Soluble in water.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H34CaN2O11
Molecular Weight (g/mol) 494.551
MDL Number MFCD00002766
InChI Key KUKRZUPDYACRAM-GXIDORRSSA-L
Synonym calcium 3-2,4-dihydroxy-3,3-dimethylbutanamido propanoate hydrate, d-pantothenic acid calcium salt hydrate, calcium hydrate dipantothenate, calcium bis 3-2,4-dihydroxy-3,3-dimethylbutanoyl amino propanoate hydrate
PubChem CID 16211719
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate;hydrate
SMILES CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.O.[Ca+2]

Specifications

Melting Point ∼138°C (decomposition)
Beilstein 3769272
Merck Index 14,7015
Solubility Information Soluble in water.
Quantity 100 g
Optical Rotation +27° (c=5 in Water)
Formula Weight 476.54
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material D-Pantothenic acid calcium salt hydrate
EINECSNumber 205-278-9
RTECSNumber RU4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.