Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantothenic Acid, Calcium Salt , MP Biomedicals, LLC

Catalog No. MP210122880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP210122880 100 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210122880 Supplier MP Biomedicals, Inc Supplier No. 0210122880
Add to Cart
Add to Cart

D-Pantothenic acid, Calcium Salt USP grade is an essential vitamin.

  • Precursor in the biosynthesis of coenzyme A
  • Production of energy
  • Catabolism of fatty acids and amino acids
  • Synthesis of fatty acids, phospholipids, sphingolipids, cholesterol, and steroid hormones
  • Synthesis of heme and the neurotransmitter acetylcholine
  • Involved in regulation of gene expression and in signal transduction
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-83-4
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
InChI Key GHOKWGTUZJEAQD-UHFFFAOYNA-N
Synonym d-pantothenic acid, pantothenic acid, vitamin b5, pantothenate, chick antidermatitis factor, +-pantothenic acid, r-pantothenate, d-+-pantothenic acid, kyselina pantothenova czech, pantothenoic acid
PubChem CID 6613
ChEBI CHEBI:46905

Specifications

Melting Point 170°C to 172°C (literature)
Color White
pH 7.2 to 8.0 (at 5% aq. soln.) (literature)
Quantity 100 g
Packaging Glass
Density 1.266 (Lit.)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.