Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantothenic acid calcium salt, USP grade, MP Biomedicals™

Catalog No. ICN10122805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10122805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10122805 Supplier MP Biomedicals, Inc Supplier No. 0210122805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity USP grade Vitamin B5, essential as a precursor in the biosynthesis of coenzyme A.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 238.3
Grade USP
Physical Form Powder
Chemical Name or Material D-Pantothenic acid calcium salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.